Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL3/MIP-1 alpha Rat anti-Mouse, Clone: 756605, R&D Systems™

Rat MonoclonalAntibody

$116.82 - $344.50

Specifications

Antigen CCL3/MIP-1 alpha
Clone 756605
Dilution Neutralization 0.125-0.75 ug/mL
Applications Neutralization
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
MAB4502100 View Documents R&D Systems
MAB4502100
100 ug Unconjugated Each for $344.50
Add to cart
 
MAB4502SP View Documents R&D Systems
MAB4502SP
25 ug Unconjugated Each for $116.82
Add to cart
 
Description

Description

<0.10 EU per 1µg of the antibody by the LAL method.
Specifications

Specifications

CCL3/MIP-1 alpha
Neutralization 0.125-0.75 ug/mL
Purified
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
E. coli-derived recombinant mouse CCL3/MIP-1 alpha Ala24-Ala92 Accession # P10855
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Detects mouse CCL3/MIP-1 alpha in direct ELISAs.
756605
Neutralization
Rat
RUO
6348
IgG1
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit