Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCL3L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596214
Description
Immunogen sequence: APLAADTPTA CCFSYTSRQI PQNFIADYFE TSSQCSKPSV IFLTKRGRQV CADPSEEWVQ KYVSDLELSA; Positive Samples: B-cell.
This gene is one of several cytokine genes that are clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins that function in inflammatory and immunoregulatory processes. The protein encoded by this gene binds to several chemokine receptors, including chemokine binding protein 2 and chemokine (C-C motif) receptor 5 (CCR5). CCR5 is a co-receptor for HIV, and binding of this protein to CCR5 inhibits HIV entry. The copy number of this gene varies among individuals, where most individuals have one to six copies, and a minority of individuals have zero or more than six copies. There are conflicting reports about copy number variation of this gene and its correlation to disease susceptibility. This record represents one of two copies that are present on the ALT_REF_LOCI_2 alternate haplotype of the GRCh38 human reference genome assembly. Alternative splicing of this gene results in multiple transcript variants.
Specifications
CCL3L1 | |
Polyclonal | |
Unconjugated | |
CCL3L1 | |
464.2; C-C motif chemokine 3-like 1; C-C motif chemokine ligand 3 like 1; CC-chemokine (CCL3); MIP1a; CCL3L1; CCL3L3; chemokine (C-C motif) ligand 3-like 1; chemokine ligand 3-like 1; D17S1718; G0/G1 switch regulatory protein 19-2; G0S19-2; LD78; LD78BETA; LD78-beta(1-70); LD78-beta(3-70); LD78-beta(5-70); macrophage inflamatory protein 1 alpha; MIP1AP; PAT 464.2; SCYA3L; SCYA3L1; small inducible cytokine A3-like 1; Small-inducible cytokine A3-like 1; Tonsillar lymphocyte LD78 beta protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6349 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16619 | |
CCL3L1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 24-93 of human CCL3L1 (NP_0662861). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction