Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL6/C10 Antibody (262016), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB487W
Description
CCL6/C10 Monoclonal antibody specifically detects CCL6/C10 in Mouse samples. It is validated for Western Blot, Neutralization, Flow Cytometry, CyTOFSpecifications
CCL6/C10 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Neutralization, Flow Cytometry, CyTOF | |
262016 | |
Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
c10,C-C motif chemokine 6,MRP,MRP-1,Scy,Scya6 | |
E. coli-derived recombinant mouse CCL6/C10, Gly22-Ala116, Accession # P27784 | |
0.5 mL | |
Chemokines and Cytokines | |
20305 | |
Store at 4C in the dark. | |
IgG2b |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction