Learn More
CCL6/C10 Rat anti-Mouse, Alexa Fluor™ 594, Clone: 11D7, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221985AF594
Catalog No. NP22198A94
Add to cart
Description
Description
CCL6/C10 Monoclonal antibody specifically detects CCL6/C10 in Mouse samples. It is validated for ELISA.
Specifications
Specifications
CCL6/C10
11D7
ELISA
Ccl6
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein CCL-6. Rat monoclonal anti mouse CCL6 P27784.
0.1 mL
Primary
20305
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse