Learn More
CCL6 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Supplier: Pepro Tech Inc 500P112BT25UG

Description
AA Sequence of recombinant protein: GLIQEIEKED RRYNPPIIHQ GFQDTSSDCC FSYATQIPCK RFIYYFPTSG GCIKPGIIFI SRRGTQVCAD PSDRRVQRCL STLKQGPRSG NKVIA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine C10 (CCL6). Anti-Murine C10 (CCL6)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine C10 (CCL6) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine C10 (CCL6) (500-P112) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine C10 (CCL6). Western Blot: To detect mC10 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mC-10 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
CCL6 Polyclonal antibody specifically detects CCL6 in Mouse samples. It is validated for ELISA, Western BlotSpecifications
CCL6 | |
Polyclonal | |
PBS with no preservative | |
P27784 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20305 | |
-20°C |
ELISA, Western Blot | |
Biotin | |
Ccl6 | |
c10; CC chemokine C10; C-C motif chemokine 6; Ccl6; CCL6(22-95); CCL6(23-95); chemokine (C-C motif) ligand 6; chemokine ligand 6; Mrp-1; Protein C10; Scay6; Scya6; small inducible cytokine A6; small-inducible cytokine A6 | |
E.coli-derived Recombinant Murine C10 (CCL6) | |
25 μg | |
Primary | |
Mouse | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.