Learn More
CCL7/MCP-3/MARC Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0156-6J48, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212122AF594
Catalog No. NP212122A94
Add to cart
Description
Description
CCL7/MCP-3/MARC Monoclonal antibody specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL7/MCP-3/MARC
MM0156-6J48
Western Blot
C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3)
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CCL7
Human MCP3
0.1 mL
Primary
6354