Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL7/MCP-3/MARC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18236720UL
Description
CCL7/MCP-3/MARC Polyclonal specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CCL7/MCP-3/MARC | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry | |
NP_006264 | |
CCL7 | |
Synthetic peptide towards MCP3. Peptide sequence TSTTCCYRFINKKIPKQRLESYRRTTSSHCPREAVIFKTKLDKEICADPT. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
PBS & 2% Sucrose. with No Preservative | |
C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3) | |
Rabbit | |
9 kDa | |
20 μL | |
Cancer | |
6354 | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction