Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL7/MCP-3/MARC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$157.00 - $482.50
Specifications
Antigen | CCL7/MCP-3/MARC |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18236720
|
Novus Biologicals
NBP18236720UL |
20 μL |
Each for $157.00
|
|
|||||
NBP182367
|
Novus Biologicals
NBP182367 |
100 μL |
Each for $482.50
|
|
|||||
Description
CCL7/MCP-3/MARC Polyclonal specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CCL7/MCP-3/MARC | |
Unconjugated | |
RUO | |
NP_006264 | |
6354 | |
Synthetic peptide towards MCP3. Peptide sequence TSTTCCYRFINKKIPKQRLESYRRTTSSHCPREAVIFKTKLDKEICADPT. | |
Primary |
Polyclonal | |
Rabbit | |
Cancer | |
C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3) | |
CCL7 | |
IgG | |
9 kDa |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title