Learn More
CCL8/MCP-2 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0157-7D27, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212124AF594
Catalog No. NP212124A94
Add to cart
Description
Description
CCL8/MCP-2 Monoclonal antibody specifically detects CCL8/MCP-2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CCL8/MCP-2
MM0157-7D27
Western Blot
C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CCL8
Human MCP2
0.1 mL
Primary
6355