Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL8/MCP-2 Antibody (RM0184-17A2), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212123AF350
Description
CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Mouse samples. It is validated for Western Blot.Specifications
CCL8/MCP-2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CCL8 | |
Mouse recombinant CCL8/MCP-2 | |
0.1 mL | |
6355 | |
Store at 4°C in the dark. |
Western Blot | |
RM0184-17A2 | |
Western Blot | |
C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction