Learn More
CCL8/MCP-2 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0184-17A2, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212123AF594
Catalog No. NP212123A94
Add to cart
Description
Description
CCL8/MCP-2 Monoclonal antibody specifically detects CCL8/MCP-2 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CCL8/MCP-2
RM0184-17A2
Western Blot
C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CCL8
Mouse MCP2
0.1 mL
Primary
6355