Learn More
CCL9 Rat anti-Mouse, Alexa Fluor 405, Clone: RM0185-28B3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212408AF405
Catalog No. NB212408405
Add to cart
Description
Description
CCL9 Monoclonal specifically detects CCL9 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CCL9
RM0185-28B3
Western Blot
Purified
CCL9
Mouse
Protein G purified
Chemokines and Cytokines
Primary
20308
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CCF18, Ccl9 chemokine (C-C motif) ligand 9, MRP-2, Scya10, Scya9
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse