Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCL9 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0185-28B3, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212408AF750
Description
CCL9 Monoclonal specifically detects CCL9 in Mouse samples. It is validated for Western Blot.Specifications
CCL9 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CCL9 | |
Mouse recombinant CCL9 | |
0.1 mL | |
20308 | |
Store at 4°C in the dark. |
Western Blot | |
RM0185-28B3 | |
Western Blot | |
CCF18, Ccl9 chemokine (C-C motif) ligand 9, MRP-2, Scya10, Scya9 | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title