Learn More
CCNB1 Mouse anti-Hamster, Human, Clone: V63, Abnova™
Mouse monoclonal antibody raised against hamster Ccnb1.
Manufacturer:  Abnova Corporation MAB12596
Catalog No. 89-933-732
Add to cart
Description
Description
Specifications
Specifications
CCNB1
V63
Mouse monoclonal antibody raised against hamster Ccnb1.
In PBS (50% glycerol, 0.09% sodium azide).
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Hamster Ccnb1
Protein G purification
RUO
Primary
Hamster, Human