Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNB1IP1 Mouse anti-Human, Clone: OTI4H3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502700

Catalog No. 50-167-3735


Add to cart

Description

Description

HEI10 is a member of the E3 ubiquitin ligase family and functions in progression of the cell cycle through G(2)/M.
Specifications

Specifications

CCNB1IP1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NPC3
C14orf18, HEI10
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57820
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI4H3
Unconjugated
CCNB1IP1
Liquid
CCNB1IP1
Full length human recombit protein of human CCNB1IP1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit