Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCND1 Mouse anti-Human, Clone: CCND1/809, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14432

Catalog No. 89-034-515


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human CCND1.

  • High specificity
  • High reactivity
Specifications

Specifications

CCND1
CCND1/809
Mouse monoclonal antibody raised against full length recombinant human CCND1.
In 10 mM PBS (0.05% BSA and 0.05% azide).
Liquid
CCND1
Recombinant protein corresponding to full length human CCND1.
Protein A/G purification
RUO
Primary
595
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by
P24385
BCL1|D11S287E|PRAD1|U21B31
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)