Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
CCND2 Mouse anti-Human, Clone: OTI1E4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806252
Catalog No. 50-166-7761
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cyclin D2 is a regulatory component of the cyclin D2-CDK4 (DC) complex that phosphorylates and inhibits members of the retinoblastoma (RB) protein family including RB1 and regulates the cell-cycle during G1/S transition. Phosphorylation of RB1 allows dissociation of the transcription factor E2F from the RB/E2F complex and the subsequent transcription of E2F target genes which are responsible for the progression through the G1 phase. Hypophosphorylates RB1 in early G1 phase. Cyclin D-CDK4 complexes are major integrators of various mitogenenic and antimitogenic signals. Also substrate for SMAD3, phosphorylating SMAD3 in a cell-cycle-dependent manner and repressing its transcriptional activity. Component of the ternary complex, cyclin D2/CDK4/CDKN1B, required for nuclear translocation and activity of the cyclin D-CDK4 complex.
Specifications
Specifications
CCND2
Monoclonal
Unconjugated
CCND2
Lyophilized
CCND2
Full length human recombit protein of human CCND2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1E4
PBS with 8% trehalose and no preservative; pH 7.3
P30279
KIAK0002, MPPH3
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
894