Learn More
Abnova™ CCNF 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000899T01
Catalog No. 89-938-507
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CCNF 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 86.57kDa
Transient overexpression cell lysate was tested with anti-CCNF antibody (Mfr. No. H00000899-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
BC012349
pCMV-CCNF full-length
100 μL
Human