Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCNJL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597552
Description
CCNJL Polyclonal Antibody for Western Blot, IHC (P), ELISA
CCNJL is a protein coding gene and gene ontology (GO) annotation includes cyclin-dependent protein kinase holoenzyme complex; cytoplasm; nucleus; cyclin-dependent protein serine/threonine kinase regulator activity; protein kinase binding; mitotic cell cycle phase transition; protein phosphorylation; regulation of cyclin-dependent protein serine/threonine kinase activity.
Specifications
CCNJL | |
Polyclonal | |
Unconjugated | |
CCNJL | |
Ccnjl; cyclin J like; cyclin J-like; cyclin-J-like protein; Gm877 | |
Rabbit | |
Protein G | |
RUO | |
380694, 79616 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q5SRT8, Q8IV13 | |
CCNJL | |
Recombinant Human Cyclin-J-like protein (315-435AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction