Learn More
CCR1 Rat anti-Mouse, Alexa Fluor™ 405, Clone: 643854R, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB5986RV100UG
Catalog No. FB5986RV100
Add to cart
This item is not returnable. View return policy
Description
Description
CCR1 Monoclonal antibody specifically detects CCR1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR1
643854R
Flow Cytometry 0.25-1 ug/10^6 cells
P51675
C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R
E. coli-derived recombinant mouse CCR1, Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281), Accession No. P51675
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse CCR1 in direct ELISAs.
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Rat
IgG2b
100 μg
Primary
1230.0
Mouse