Learn More
Mouse CCR1 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB5986RR100UG
Catalog No. FB5986RR100
Add to Cart
This item is not returnable. View return policy
Description
Description
CCR1 Monoclonal specifically detects CCR1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR1
Monoclonal
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R
E. coli-derived recombinant mouse CCR1, Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281), Accession No. P51675
100 μg
1230.0
Mouse
Supernatant
Flow Cytometry
643854R
Flow Cytometry 0.25-1 ug/10^6 cells
P51675
Rat
Protein A or G purified from cell culture supernatant
Primary
Detects mouse CCR1 in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2b
Product Suggestions
Product Suggestions