Learn More
CCR10/GPR2 Rat anti-Mouse, Alexa Fluor 647, Clone: 248918R, R&D Systems
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB2815RR100UG
Catalog No. FB2815RR100
Add to cart
This item is not returnable. View return policy
Description
Description
CCR10/GPR2 Monoclonal antibody specifically detects CCR10/GPR2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR10/GPR2
248918R
Flow Cytometry 0.25-1 μg/106 cells
NP_031747
CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2
C6 rat glioma cell line transfected with mouse CCR10, Met1-Asn362, Accession No. NP_031747
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse CCR10 in direct ELISAs.
Flow Cytometry
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Rat
IgG2a
100 μg
Primary
2826.0
Mouse