Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR10 Mouse anti-Human, BUV395, Clone: 1B5, BD Horizon™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 565322

Catalog No. BDB565322


Only null left
Add to Cart

Description

Description

CCR10 Monoclonal antibody specifically detects CCR10 in Human samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCR10
Monoclonal
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
25 μg
Cell Biology
Human
Purified
Flow Cytometry
1B5
Brilliant Ultraviolet 395
C-C CKR-10, C-C CKR-10, G-protein coupled receptor 2, CCR-10, GPR2
Human CCR10 Transfected Cell Line
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity and unconjugated antibody and free BD Horizon™ BUV395 were removed. chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Reseach Use Only.