Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR11 Goat anti-Human, Mouse, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
$176.85
Specifications
Antigen | CCR11 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500, ELISA 1:37500, Immunohistochemistry 1:125, Immunohistochemistry-Paraffin 1:125 |
Applications | ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017798
|
Novus Biologicals
NB1007050.025MG |
0.025 mg |
Each for $176.85
|
|
NB100705
|
Novus Biologicals
NB100705 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
CCR11 Polyclonal specifically detects CCR11 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR11 | |
Western Blot 1:500, ELISA 1:37500, Immunohistochemistry 1:125, Immunohistochemistry-Paraffin 1:125 | |
Polyclonal | |
Goat | |
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide | |
51554 | |
Synthetic peptide: EQNQSTDYYYEENEMNGTYDYSQYELIC, corresponding to amino acids 4-31 of Human CCR11. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse | |
CC chemokine receptor-like 1, C-C CKR-11, CCBP2CCX CKR, CCR11CCR10, chemokine (C-C motif) receptor-like 1, orphan seven-transmembrane receptor, chemokine related, PPR1cc motif, receptor-like protein 1, VSHK1CKR-11 | |
ACKR4 | |
IgG | |
Affinity Purified | |
Peptide sequence is < 50 % identical to other human chemokine receptors in this region. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title