Learn More
CCR2 (CD192) Mouse anti-Human, Brilliant Violet 510, Clone: LS132.1D9, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 747851
Catalog No. BDB747851
Add to cart
Description
Description
CCR2 (CD192) Monoclonal antibody specifically detects CCR2 (CD192) in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR2 (CD192)
LS132.1D9 (also known as 1D9)
Brilliant Violet 510
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Human
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG2a, κ
RUO
Primary
CCR2 (CD192)