Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR2 Goat anti-Human, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
$153.27 - $436.00
Specifications
Antigen | CCR2 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500, ELISA 1:30000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
CCR2 Polyclonal specifically detects CCR2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR2 | |
Western Blot 1:500, ELISA 1:30000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
Polyclonal | |
Goat | |
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
10mM KHPO4 and 0.14M NaCl with 0.1% Sodium Azide | |
729230 | |
Synthetic peptide C-G17EEVTTFFDYDYGAPCHKFDVKQ39 corresponding to the N-terminal extracellular domain of human CCR2. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
CCR2 | |
IgG | |
Affinity Purified | |
Peptide sequence is < 50 % identical to other human chemokine receptors in this region. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title