Learn More
CCR2 Mouse anti-Rat, Alexa Fluor 350, Clone: 890231, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8368U100UG
Catalog No. FAB8368U100
Add to cart
This item is not returnable. View return policy
Description
Description
CCR2 Monoclonal antibody specifically detects CCR2 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR2
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
O55193
CCR2
NS0 mouse myeloma cell line transfected with rat CCR2 Accession # O55193
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains rat CCR2 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
890231
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
Mouse
IgG2b
RUO
Primary
729230
Rat
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)