Learn More
CCR2 Mouse anti-Rat, Alexa Fluor™ 350, Clone: 890231R, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8368RU100UG
Catalog No. FAB8368RU10
Add to cart
This item is not returnable. View return policy
Description
Description
CCR2 Monoclonal antibody specifically detects CCR2 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR2
890231R
Flow Cytometry 0.25-1 μg/mL
O55193
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
NS0 mouse myeloma cell line transfected with rat CCR2, Accession No. O55193
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects rat CCR2 in direct ELISAs.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
729230.0
Rat