Learn More
CCR2 Mouse anti-Rat, Alexa Fluor 405, Clone-890231, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8368V025UG
Catalog No. FAB8368V025
Add to cart
This item is not returnable. View return policy
Description
Description
Flow Cytometry: Please use 0.25-1 ug of conjugated antibody per 10e6 cells.
Specifications
Specifications
CCR2
890231
Alexa Fluor 405
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
Mouse
IgG2b
25 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Stains rat CCR2 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
0.2 mg/mL
Flow Cytometry 1.0 ug/10^6 cells
O55193
CCR2
NS0 mouse myeloma cell line transfected with rat CCR2 Accession # O55193
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
729230
Rat