Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR2 Mouse anti-Rat, Alexa Fluor™ 647, Clone: 890231R, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8368RR100UG
This item is not returnable.
View return policy
Description
CCR2 Monoclonal specifically detects CCR2 in Rat samples. It is validated for Flow Cytometry.Specifications
CCR2 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
NS0 mouse myeloma cell line transfected with rat CCR2, Accession No. O55193 | |
100 μg | |
729230.0 | |
Rat | |
Supernatant |
Flow Cytometry | |
890231R | |
Flow Cytometry 0.25-1 μg/mL | |
O55193 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects rat CCR2 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title