Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR3, Rabbit anti-Human, Clone: 5E8-G9-B4, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen CCR3
Clone 5E8-G9-B4
Applications Flow Cytometry
Host Species Rabbit
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB014770
View Documents
Novus Biologicals
NBP2810460.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB014771
View Documents
Novus Biologicals
NBP2810460.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

CCR3 Monoclonal antibody specifically detects CCR3 in Human Samples. It is validated for Flow Cytometry.
Specifications

Specifications

CCR3
Flow Cytometry
Human
Primary
5E8-G9-B4
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit