Learn More
CCR4 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0064-9G12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212129AF532
Catalog No. NP212129A32
Add to cart
Description
Description
CCR4 Monoclonal antibody specifically detects CCR4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR4
MM0064-9G12
Flow Cytometry
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CCR4
Human CCR4
0.1 mL
Primary
1233