Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR4 Antibody (MM0064-9G12), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212129AF594
Description
CCR4 Monoclonal specifically detects CCR4 in Human samples. It is validated for Flow Cytometry.Specifications
CCR4 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CCR4 | |
Human CCR4 transfected 293 cells | |
Primary | |
Detects human CCR4, not other CCRs | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry (Frozen) | |
MM0064-9G12 | |
Flow Cytometry, Immunohistochemistry-Frozen | |
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
Mouse | |
0.1 mL | |
1233 | |
Human | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction