Learn More
CCR4 Mouse anti-Human, Alexa Fluor 700, Clone: 205410R, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB1567RN100UG
Catalog No. FB1567RN100
Add to cart
This item is not returnable. View return policy
Description
Description
CCR4 Monoclonal antibody specifically detects CCR4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR4
205410R
Flow Cytometry 0.25-1 μg/106 cells
P51679
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5
Human CCR4 transfectants, Met1-Leu360, Accession No. P51679
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human CCR4 in direct ELISAs
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
1233.0
Human