Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR4 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Supplier:  Thermo Scientific PA129771

Catalog No. PIPA129771


Add to Cart

Description

Description

A suggested positive control for this product is mouse spleen cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CCR4 Polyclonal antibody specifically detects CCR4 in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCR4
Polyclonal
Unconjugated
Ccr4
C-C chemokine receptor 4; C-C chemokine receptor type 4; C-C CKR-4; C-C motif chemokine receptor 4; CC-CKR-4; Ccr4; CCR-4; CD194; chemokine (C-C motif) receptor 4; chemokine (C-C) receptor 4; CHEMR1; ChemR13; CKR4; Cmkbr4; fusin; HGCN:14099; K5-5; LESTR; leukocyte-expressed seven-transmembrane-domain receptor; MGC88293; Sdf1r
Goat
Antigen affinity chromatography
RUO
12773
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.1% sodium azide
P51680
Ccr4
Synthetic peptide corresponding to residues D(8)TTQDETVYNSYYFYESMP(26)C of Mouse CCR4
50 μg
Primary
Mouse
Antibody
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit