Learn More
CCR5 Mouse anti-Human, Alexa Fluor 700, Clone: CTC5R, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB1802RN100UG
Catalog No. FB1802RN100
Add to cart
This item is not returnable. View return policy
Description
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR5
CTC5R
Flow Cytometry 0.25-1 μg/106 cells
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
1234.0
Human
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
CHO Chinese hamster ovary cell line transfected with human CCR5
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human CCR5 in direct Flow cytometry.