Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR5 Mouse anti-Human, PE-Cyanine7, Clone: 45523.111, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB12010397PECY7

Catalog No. N2010397PC7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CCR5
45523.111
Flow Cytometry
Conjugated, Purified
CCR5
Human CCR5 transfected NSO mouse myeloma cells.
Protein G purified
Alzheimers Research, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction
Primary
1234.0
Human
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
The antibody reacts with human CCR-5 transfectant cells and not the parent cell line. The antibody shows no cross-reactivity with CCR-1, CCR-2 or CCR-3. This antibody can be used to detect CCR-5 present on stimulated human PBMCs that have been fixed with 2% formaldehyde, but not on unfixed cells.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit