Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR5 Rat anti-Human, PE-Cyanine7, Clone: HEK/1/85a/7a, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP260258PECY7

Catalog No. N260258PEC7


Add to cart

Description

Description

CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CCR5
HEK/1/85a/7a
Flow Cytometry
Conjugated, Purified
CCR5
CHO cells transfected with human CCR5
Protein G purified
Alzheimers Research, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction
Primary
1234.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
Rat
IgG2a Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit