Learn More
CCR5 Rat anti-Mouse, Alexa Fluor™ 647, Clone: 225307, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB18041R100UG
Catalog No. FB18041R100
Add to cart
This item is not returnable. View return policy
Description
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR5
225307
Flow Cytometry 0.25-1 ug/10^6 cells
P51682
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
C6 rat glioma cell line transfected with mouse CCR5, Accession No. P51682
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse CCR5 transfectants but not the parental cell line in Flow Cytometry and Western blots.
Flow Cytometry
Alexa Fluor™ 647
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Rat
IgG2b
100 μg
Primary
1234.0
Mouse