Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCR5 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB18041S100UG
This item is not returnable.
View return policy
Description
CCR5 Monoclonal specifically detects CCR5 in Mouse samples. It is validated for Flow Cytometry.Specifications
CCR5 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide | |
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
C6 rat glioma cell line transfected with mouse CCR5, Accession No. P51682 | |
Primary | |
Detects mouse CCR5 transfectants but not the parental cell line in Flow Cytometry and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
Flow Cytometry | |
225307 | |
Flow Cytometry 0.25-1 μg/106 cells | |
P51682 | |
Rat | |
100 μg | |
1234 | |
Mouse | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction