Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR6 Mouse anti-Rat, Alexa Fluor 488, Clone: 876515, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8320G100UG
This item is not returnable.
View return policy
Description
CCR6 Monoclonal specifically detects CCR6 in Rat samples. It is validated for Flow Cytometry.Specifications
CCR6 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q5BK58 | |
CCR6 | |
NSO mouse myeloma cell line transfected with rat CCR6 Met1-Met366 Accession # Q5BK58 | |
RUO | |
1235 | |
Rat | |
IgG2b |
Flow Cytometry | |
876515 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22 | |
Mouse | |
100 μg | |
Primary | |
Detects rat CCR6 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)