Learn More
CCR6 Mouse anti-Rat, Alexa Fluor 750, Clone: 876515, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8320S100UG
Catalog No. FAB8320S100
Add to cart
This item is not returnable. View return policy
Description
Description
CCR6 Monoclonal antibody specifically detects CCR6 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
CCR6
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q5BK58
CCR6
NSO mouse myeloma cell line transfected with rat CCR6 Met1-Met366 Accession # Q5BK58
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects rat CCR6 in direct ELISAs.
Flow Cytometry
876515
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22
Mouse
IgG2b
RUO
Primary
1235
Rat
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)