Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR6 Rat anti-Mouse, Alexa Fluor 488, Clone: 140706R, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB590RG100UG
This item is not returnable.
View return policy
Description
CCR6 Monoclonal specifically detects CCR6 in Mouse samples. It is validated for Flow Cytometry.Specifications
CCR6 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22 | |
RBL-1 rat basophilic leukemia cell line transfected with mouse CCR6 Met1-Met367 Accession # O54689 | |
100 μg | |
1235 | |
Mouse | |
Supernatant |
Flow Cytometry | |
140706R | |
Flow Cytometry 0.25-1μg/106 cells | |
O54689 | |
Rat | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects mouse CCR6-transfected rat RBL cells but not the parent cell line or cells transfected with mouse CCR5. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title