Learn More
CCR6 Rat anti-Mouse, Alexa Fluor™ 532, Clone: 18B9E6, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225224AF532
Catalog No. NP22224A32
Add to cart
Description
Description
CCR6 Monoclonal antibody specifically detects CCR6 in Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CCR6
18B9E6
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22
Rat
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CCR6
A recombinant protein consisting of the external domains of mouse CD196 (CCR6) was used as the immunogen for the antibody.
0.1 mL
Primary
1235