Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR6 Rat anti-Human, Mouse, Alexa Fluor 594, Clone: 18B9E6, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP225224AF594
Description
CCR6 Monoclonal specifically detects CCR6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR6 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CCR6 | |
A recombinant protein consisting of the external domains of mouse CD196 (CCR6) was used as the immunogen for the antibody. | |
Primary | |
Human, Mouse | |
IgG2a κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
18B9E6 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22 | |
Rat | |
0.1 mL | |
1235 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title