Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR7 (CD197) Mouse anti-Human , APC, Clone: 2-L1-A, BD Biosciences™

Mouse Monoclonal Antibody

$171.60 - $326.50

Specifications

Antigen CCR7 (CD197)
Clone 2-L1-A
Applications Flow Cytometry
Conjugate APC
Format Conjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566763
View Documents
BD Biosciences
566763
25 Tests
Each for $171.60
Add to cart
 
BDB566762
View Documents
BD Biosciences
566762
100 Tests
Each for $326.50
Add to cart
 
Description

Description

CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CCR7 (CD197)
Flow Cytometry
Conjugated
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CCR7 Transfected Cell Line
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed.
2-L1-A
APC
Mouse
Human
BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor
Mouse BALB/c IgG1, κ
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit