Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human , BV510, Clone: 2-L1-A, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566760
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow CytometrySpecifications
CCR7 (CD197) | |
Monoclonal | |
Brilliant Violet 510 | |
BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor | |
Human CCR7 Transfected Cell Line | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
2-L1-A | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV510 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV510 were removed. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction