Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR7 Rat anti-Human, Phycoerythrin, Clone: 3D12, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143332PE

Catalog No. NBP143332PE


Add to cart

Description

Description

CCR7 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin).
Specifications

Specifications

CCR7
3D12
Flow Cytometry
Purified
CCR7
The immunogen for this antibody was CCR7.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor
Rat
IgG2a Kappa
0.1 ml
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary
1236
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit