Learn More
CCR8 Rabbit anti-Mouse, Alexa Fluor 700, Clone: 1055C, R&D Systems™
Rabbit MonoclonalAntibody
Manufacturer:  R&D Systems FAB8324N100UG
Catalog No. FAB8324N100
Add to cart
This item is not returnable. View return policy
Description
Description
Recombinant Monoclonal Antibody.
Specifications
Specifications
CCR8
Monoclonal
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1
HEK293 human embryonic kidney cell line transfected with mouse CCR8 Accession # P56484
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains mouse CCR8 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
1055C
Flow Cytometry 0.25-1 ug/10^6 cells
P56484
Rabbit
IgG
RUO
Primary
1237
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)