Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR9 Goat anti-Human, Mouse, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 1 publication
$153.25 - $436.00
Specifications
Antigen | CCR9 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:100, Immunohistochemistry 1:125, Immunohistochemistry-Paraffin 1:125 |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
CCR9 Polyclonal specifically detects CCR9 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR9 | |
Flow Cytometry 1:100, Immunohistochemistry 1:125, Immunohistochemistry-Paraffin 1:125 | |
Polyclonal | |
Goat | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
10mM KHPO4 and 0.14M NaCl with 0.1% Sodium Azide | |
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
CCR9 | |
IgG | |
Affinity Purified | |
Peptide sequence is < 50 % identical to other mouse chemokine receptors in this region. |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
Q9WUT7 | |
10803 | |
Synthetic peptide: IPGMFDDFSYDSTASTDDYMNLNFSSFF, corresponding to amino acids 10-37 of Mouse CCR9. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title